Assay Details
Target Gene Details
Entrez Gene ID: | 23246 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
- |
Location: |
Chr.8:144262046-144291438 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOP1 | NM_015201.4 | NP_056016.1 | ||
AB060694.1 | BAB70666.1 | |||
AK024840.1 | ||||
AK290677.1 | ||||
AK293719.1 | ||||
AK315612.1 | ||||
BC001086.2 | AAH01086.2 | |||
BC005160.2 | AAH05160.2 | |||
BC007274.1 | AAH07274.1 | |||
BC013787.1 | AAH13787.1 | |||
BC013980.2 | AAH13980.1 | |||
BC017674.2 | AAH17674.1 | |||
BQ711345.1 | ||||
D50914.1 | BAA09473.1 | |||
DA510177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12434n54 | Chr.8:144270080 - 144516801 on Build GRCh38 | Loss | FOXH1 PPP1R16A DGAT1 RECQL4 MIR6849 SLC39A4 MIR1234 HSF1 TONSL-AS1 TMEM249 MIR6848 LOC101928902 CYHR1 MIR939 FBXL6 BOP1 SCRT1 VPS28 MIR6893 CPSF1 TONSL GPT MFSD3 LOC101928953 KIFC2 SLC52A2 ADCK5 |
nsv470249 | Chr.8:143889098 - 144519234 on Build GRCh38 | Loss | FOXH1 PARP10 EXOSC4 PPP1R16A DGAT1 LRRC14 SMPD5 RECQL4 MIR6849 SLC39A4 MIR1234 HSF1 MIR6846 TONSL-AS1 TMEM249 MIR6847 MIR6848 SPATC1 LOC101928902 CYHR1 MAF1 CYC1 MIR939 SHARPIN FBXL6 BOP1 GRINA SCRT1 VPS28 MIR6893 OPLAH CPSF1 MIR661 SCX MIR7112 TONSL GPT MFSD3 HGH1 LOC101928953 PLEC WDR97 GPAA1 MROH1 KIFC2 SLC52A2 ADCK5 |
nsv515809 | Chr.8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | FOXH1 PARP10 EXOSC4 PPP1R16A DGAT1 LRRC14 SMPD5 RECQL4 MIR6849 SLC39A4 MIR1234 HSF1 MIR6846 TONSL-AS1 TMEM249 MIR6847 MIR6848 SPATC1 LOC101928902 CYHR1 MAF1 CYC1 MIR939 SHARPIN FBXL6 BOP1 GRINA SCRT1 VPS28 MIR6893 OPLAH CPSF1 MIR661 SCX MIR7112 TONSL GPT MFSD3 HGH1 LOC101928953 PLEC WDR97 GPAA1 MROH1 KIFC2 SLC52A2 ADCK5 |
nsv1025071 | Chr.8:144044224 - 144300847 on Build GRCh38 | Gain | MAF1 CYC1 SHARPIN EXOSC4 BOP1 SMPD5 OPLAH SCX HSF1 MIR6846 MIR7112 HGH1 WDR97 GPAA1 MIR6847 MROH1 SPATC1 |
nsv428208 | Chr.8:144041826 - 144722291 on Build GRCh38 | Gain | FOXH1 EXOSC4 LRRC24 PPP1R16A DGAT1 LRRC14 SMPD5 C8orf82 RECQL4 MIR6849 SLC39A4 MIR1234 HSF1 MIR6846 TONSL-AS1 TMEM249 ZNF251 MIR6847 MIR6848 SPATC1 LOC101928902 CYHR1 MAF1 CYC1 MIR939 SHARPIN FBXL6 BOP1 SCRT1 VPS28 MIR6893 OPLAH CPSF1 SCX MIR7112 TONSL GPT MFSD3 HGH1 LOC101928953 ARHGAP39 WDR97 GPAA1 MROH1 KIFC2 SLC52A2 ADCK5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |