Assay Details
Target Gene Details
Entrez Gene ID: | 528 |
Gene Name: | ATPase H+ transporting V1 subunit C1 |
Gene Aliases: |
ATP6C, ATP6D, VATC, Vma5 |
Location: |
Chr.8:103021020-103073057 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V1C1 | NM_001695.4 | NP_001686.1 | ||
AK000064.1 | ||||
AK298617.1 | ||||
AK308632.1 | ||||
AL707882.1 | ||||
AU137773.2 | ||||
AV649881.2 | ||||
BC010960.1 | AAH10960.1 | |||
BX648079.1 | ||||
X69151.1 | CAA48903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss | ATP6V1C1 KLF10 AZIN1-AS1 UBR5 LOC401472 AZIN1 LOC101927245 ODF1 LOC107986873 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |