Assay Details
Target Gene Details
Entrez Gene ID: | 91782 |
Gene Name: | charged multivesicular body protein 7 |
Gene Aliases: |
- |
Location: |
Chr.8:23243637-23262000 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHMP7 | NM_001317899.1 | NP_001304828.1 | ||
NM_152272.4 | NP_689485.1 | |||
XM_006716415.2 | XP_006716478.1 | |||
XM_017013961.1 | XP_016869450.1 | |||
XM_017013962.1 | XP_016869451.1 | |||
XM_017013963.1 | XP_016869452.1 | |||
XM_017013964.1 | XP_016869453.1 | |||
XM_017013965.1 | XP_016869454.1 | |||
AK021776.1 | ||||
AK092500.1 | ||||
AK097202.1 | ||||
AK296592.1 | ||||
AK315664.1 | ||||
AL833843.1 | CAD38703.2 | |||
BC004344.1 | AAH04344.1 | |||
BC019110.1 | AAH19110.1 | |||
BC042050.1 | AAH42050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1027718 | Chr.8:22805950 - 23258334 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1017570 | Chr.8:22969562 - 23544783 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)