Assay Details
Target Gene Details
Entrez Gene ID: | 23246 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
- |
Location: |
Chr.8:144262046-144291438 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOP1 | NM_015201.4 | NP_056016.1 | ||
AB060694.1 | BAB70666.1 | |||
AK024840.1 | ||||
AK290677.1 | ||||
AK293719.1 | ||||
AK315612.1 | ||||
BC001086.2 | AAH01086.2 | |||
BC005160.2 | AAH05160.2 | |||
BC007274.1 | AAH07274.1 | |||
BC013787.1 | AAH13787.1 | |||
BC013980.2 | AAH13980.1 | |||
BC017674.2 | AAH17674.1 | |||
BQ711345.1 | ||||
D50914.1 | BAA09473.1 | |||
DA510177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12434n54 | Chr.8:144270080 - 144516801 on Build GRCh38 | Loss | MIR6849 FBXL6 MIR939 DGAT1 TONSL-AS1 MFSD3 LOC101928953 MIR6848 GPT PPP1R16A MIR1234 FOXH1 BOP1 MIR6893 HSF1 TMEM249 VPS28 CYHR1 KIFC2 LOC101928902 ADCK5 SLC52A2 SLC39A4 TONSL CPSF1 SCRT1 RECQL4 |
nsv470249 | Chr.8:143889098 - 144519234 on Build GRCh38 | Loss | MIR6849 FBXL6 WDR97 MIR939 PARP10 DGAT1 TONSL-AS1 MFSD3 SCX MIR6846 LOC101928953 MAF1 MIR6848 OPLAH GPT MIR7112 PPP1R16A MIR1234 FOXH1 BOP1 MROH1 MIR6847 MIR6893 HSF1 SHARPIN TMEM249 PLEC MIR661 HGH1 GRINA EXOSC4 VPS28 CYHR1 KIFC2 LOC101928902 ADCK5 CYC1 SLC52A2 SLC39A4 TONSL CPSF1 SCRT1 SPATC1 LRRC14 SMPD5 RECQL4 GPAA1 |
nsv515809 | Chr.8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | MIR6849 FBXL6 WDR97 MIR939 PARP10 DGAT1 TONSL-AS1 MFSD3 SCX MIR6846 LOC101928953 MAF1 MIR6848 OPLAH GPT MIR7112 PPP1R16A MIR1234 FOXH1 BOP1 MROH1 MIR6847 MIR6893 HSF1 SHARPIN TMEM249 PLEC MIR661 HGH1 GRINA EXOSC4 VPS28 CYHR1 KIFC2 LOC101928902 ADCK5 CYC1 SLC52A2 SLC39A4 TONSL CPSF1 SCRT1 SPATC1 LRRC14 SMPD5 RECQL4 GPAA1 |
esv2738055 | Chr.8:144277782 - 144278019 on Build GRCh38 | Deletion | BOP1 |
nsv1025071 | Chr.8:144044224 - 144300847 on Build GRCh38 | Gain | BOP1 MROH1 WDR97 MIR6847 HSF1 SHARPIN SCX HGH1 EXOSC4 MIR6846 MAF1 CYC1 OPLAH MIR7112 SPATC1 SMPD5 GPAA1 |
nsv428208 | Chr.8:144041826 - 144722291 on Build GRCh38 | Gain | MIR6849 FBXL6 WDR97 MIR939 DGAT1 TONSL-AS1 MFSD3 SCX ARHGAP39 MIR6846 LOC101928953 MAF1 MIR6848 OPLAH GPT MIR7112 PPP1R16A MIR1234 FOXH1 LRRC24 BOP1 MROH1 MIR6847 ZNF251 MIR6893 HSF1 SHARPIN TMEM249 HGH1 EXOSC4 VPS28 CYHR1 KIFC2 LOC101928902 ADCK5 CYC1 C8orf82 SLC52A2 SLC39A4 TONSL CPSF1 SCRT1 SPATC1 LRRC14 SMPD5 RECQL4 GPAA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |