Assay Details
Target Gene Details
Entrez Gene ID: | 23516 |
Gene Name: | solute carrier family 39 member 14 |
Gene Aliases: |
HMNDYT2, LZT-Hs4, NET34, ZIP14, cig19 |
Location: |
Chr.8:22367249-22434129 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC39A14 | NM_001128431.2 | NP_001121903.1 | ||
NM_001135153.1 | NP_001128625.1 | |||
NM_001135154.1 | NP_001128626.1 | |||
NM_015359.4 | NP_056174.2 | |||
XM_005273465.2 | XP_005273522.1 | |||
XM_006716323.2 | XP_006716386.1 | |||
XM_006716324.2 | XP_006716387.1 | |||
XM_011544478.2 | XP_011542780.1 | |||
XM_017013292.1 | XP_016868781.1 | |||
XM_017013293.1 | XP_016868782.1 | |||
XM_017013294.1 | XP_016868783.1 | |||
AK172810.1 | BAD18780.1 | |||
AK295807.1 | ||||
BC015770.1 | AAH15770.1 | |||
D31887.1 | ||||
DC323208.1 | ||||
DC423882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6116 | Chr.8:22368575 - 22399366 on Build GRCh38 | Insertion |
![]() |
nsv831266 | Chr.8:22315953 - 22476033 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv518646 | Chr.8:22296620 - 22520605 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2764071 | Chr.8:22342635 - 22519221 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)