Assay Details
Target Gene Details
Entrez Gene ID: | 4023 |
Gene Name: | lipoprotein lipase |
Gene Aliases: |
HDLCQ11, LIPD |
Location: |
Chr.8:19939071-19967259 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LPL | NM_000237.2 | NP_000228.1 | ||
AK092286.1 | ||||
AK222464.1 | BAD96184.1 | |||
AK290290.1 | ||||
AK297164.1 | ||||
AK312311.1 | ||||
BC011353.1 | AAH11353.1 | |||
BT006726.1 | AAP35372.1 | |||
CR457054.1 | CAG33335.1 | |||
DA485210.1 | ||||
M15856.1 | AAB59536.1 | |||
X14390.1 | CAA32564.1 | |||
X54516.1 | CAA38372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3616553 | Chr.8:19911549 - 20084901 on Build GRCh38 | Gain | LPL |
esv32767 | Chr.8:19882678 - 20015571 on Build GRCh38 | Loss | LPL |
nsv610759 | Chr.8:19704486 - 20211331 on Build GRCh38 | Gain | INTS10 LPL CSGALNACT1 SLC18A1 ATP6V1B2 |
nsv522185 | Chr.8:19097292 - 20497802 on Build GRCh38 | Gain | LOC100128993 INTS10 LPL LZTS1-AS1 SH2D4A CSGALNACT1 LZTS1 SLC18A1 ATP6V1B2 |
esv3891372 | Chr.8:19744844 - 20118728 on Build GRCh38 | Gain | INTS10 LPL CSGALNACT1 |
nsv1033511 | Chr.8:19893788 - 20767580 on Build GRCh38 | Gain | LPL LZTS1-AS1 LZTS1 SLC18A1 ATP6V1B2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |