Assay Details
Target Gene Details
Entrez Gene ID: | 23246 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
- |
Location: |
Chr.8:144262046-144291438 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOP1 | NM_015201.4 | NP_056016.1 | ||
AB060694.1 | BAB70666.1 | |||
AK024840.1 | ||||
AK290677.1 | ||||
AK293719.1 | ||||
AK315612.1 | ||||
BC001086.2 | AAH01086.2 | |||
BC005160.2 | AAH05160.2 | |||
BC007274.1 | AAH07274.1 | |||
BC013787.1 | AAH13787.1 | |||
BC013980.2 | AAH13980.1 | |||
BC017674.2 | AAH17674.1 | |||
BQ711345.1 | ||||
D50914.1 | BAA09473.1 | |||
DA510177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12434n54 | Chr.8:144270080 - 144516801 on Build GRCh38 | Loss | MIR1234 TONSL-AS1 ADCK5 MFSD3 FBXL6 DGAT1 HSF1 BOP1 PPP1R16A CYHR1 SLC39A4 TMEM249 MIR6848 KIFC2 CPSF1 MIR6893 FOXH1 TONSL SCRT1 MIR939 RECQL4 MIR6849 GPT LOC101928902 SLC52A2 VPS28 LOC101928953 |
nsv470249 | Chr.8:143889098 - 144519234 on Build GRCh38 | Loss | LRRC14 MIR1234 TONSL-AS1 ADCK5 MFSD3 FBXL6 DGAT1 SCX HSF1 SHARPIN BOP1 MROH1 PPP1R16A CYHR1 SLC39A4 SMPD5 EXOSC4 MIR6847 MIR661 TMEM249 MIR6848 KIFC2 CPSF1 MIR6893 FOXH1 TONSL SCRT1 HGH1 MIR939 CYC1 MAF1 RECQL4 MIR6849 MIR7112 OPLAH GPAA1 GPT MIR6846 LOC101928902 SLC52A2 PARP10 GRINA VPS28 WDR97 LOC101928953 SPATC1 PLEC |
esv2253218 | Chr.8:144278897 - 144279338 on Build GRCh38 | Deletion | BOP1 |
esv2738057 | Chr.8:144278710 - 144279406 on Build GRCh38 | Deletion | BOP1 |
nsv515809 | Chr.8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | LRRC14 MIR1234 TONSL-AS1 ADCK5 MFSD3 FBXL6 DGAT1 SCX HSF1 SHARPIN BOP1 MROH1 PPP1R16A CYHR1 SLC39A4 SMPD5 EXOSC4 MIR6847 MIR661 TMEM249 MIR6848 KIFC2 CPSF1 MIR6893 FOXH1 TONSL SCRT1 HGH1 MIR939 CYC1 MAF1 RECQL4 MIR6849 MIR7112 OPLAH GPAA1 GPT MIR6846 LOC101928902 SLC52A2 PARP10 GRINA VPS28 WDR97 LOC101928953 SPATC1 PLEC |
nsv1025071 | Chr.8:144044224 - 144300847 on Build GRCh38 | Gain | HGH1 CYC1 MAF1 MIR7112 SCX HSF1 SHARPIN BOP1 OPLAH GPAA1 MROH1 MIR6846 SMPD5 WDR97 EXOSC4 MIR6847 SPATC1 |
nsv428208 | Chr.8:144041826 - 144722291 on Build GRCh38 | Gain | LRRC14 MIR1234 TONSL-AS1 ADCK5 MFSD3 ZNF251 FBXL6 DGAT1 SCX HSF1 SHARPIN BOP1 MROH1 PPP1R16A CYHR1 SLC39A4 SMPD5 EXOSC4 MIR6847 TMEM249 MIR6848 KIFC2 CPSF1 MIR6893 FOXH1 C8orf82 TONSL SCRT1 HGH1 MIR939 LRRC24 CYC1 MAF1 RECQL4 MIR6849 MIR7112 OPLAH GPAA1 GPT MIR6846 LOC101928902 ARHGAP39 SLC52A2 VPS28 WDR97 LOC101928953 SPATC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |