Assay Details
Target Gene Details
Entrez Gene ID: | 100507530 |
Gene Name: | MCPH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:6615603-6708209 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MCPH1-AS1 | NR_125386.1 | |||
BX374425.2 | ||||
DA281269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1021684 | Chr.8:6492570 - 6951933 on Build GRCh38 | Loss | MIR8055 ANGPT2 AGPAT5 MIR4659A MIR4659B MCPH1 DEFB1 DEFA4 XKR5 MCPH1-AS1 GS1-24F4.2 DEFA8P DEFA6 |
nsv610004 | Chr.8:6544845 - 6915391 on Build GRCh38 | Gain | MIR8055 XKR5 MCPH1-AS1 GS1-24F4.2 ANGPT2 AGPAT5 MIR4659A MIR4659B MCPH1 DEFB1 |
nsv1025976 | Chr.8:6615427 - 6829450 on Build GRCh38 | Loss | MIR8055 XKR5 MCPH1-AS1 AGPAT5 MIR4659A MIR4659B MCPH1 |
nsv524670 | Chr.8:6643023 - 6682466 on Build GRCh38 | Loss | MCPH1-AS1 MCPH1 |
esv2422383 | Chr.8:6504868 - 6924071 on Build GRCh38 | Duplication | MIR8055 XKR5 MCPH1-AS1 GS1-24F4.2 ANGPT2 AGPAT5 MIR4659A MIR4659B MCPH1 DEFB1 |
nsv1024024 | Chr.8:6633144 - 6698037 on Build GRCh38 | Gain | MCPH1-AS1 MCPH1 |