Assay Details
Target Gene Details
Entrez Gene ID: | 81628 |
Gene Name: | TSC22 domain family member 4 |
Gene Aliases: |
THG-1, THG1, TILZ2 |
Location: |
Chr.7:100466519-100479279 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D4 | NM_001303043.1 | NP_001289972.1 | ||
NM_030935.4 | NP_112197.1 | |||
NR_130118.1 | ||||
NR_130119.1 | ||||
AJ133115.1 | CAB43491.1 | |||
AK296580.1 | ||||
AK297702.1 | ||||
AK314824.1 | ||||
BC001486.2 | AAH01486.1 | |||
BC001966.1 | AAH01966.1 | |||
BC002972.1 | AAH02972.1 | |||
BC010406.2 | AAH10406.1 | |||
BX341171.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | CLDN15 MIR6840 PCOLCE LAMTOR4 ZNHIT1 PCOLCE-AS1 PPP1R35 ZSCAN21 AZGP1P1 GPC2 PLOD3 ZAN MIR4658 ACHE MIR106B MOGAT3 LOC102724094 TFR2 MCM7 MEPCE STAG3L5P VGF NYAP1 SERPINE1 PILRB SPDYE3 NAT16 ZCWPW1 SRRT TSC22D4 LRCH4 PVRIG PVRIG2P POP7 UFSP1 GATS AGFG2 AP4M1 TAF6 MIR4653 MIR25 SAP25 ZNF3 TRIM56 C7orf61 ZKSCAN1 EPHB4 AZGP1 SLC12A9 C7orf43 MUC12 STAG3L5P-PVRIG2P-PILRB ZASP MUC3A COPS6 EPO PILRA MIR6875 CNPY4 STAG3 MOSPD3 GIGYF1 PMS2P1 AP1S1 MIR93 FBXO24 TRIP6 LOC105375429 GAL3ST4 MUC17 ACTL6B GNB2 FIS1 MBLAC1 |
nsv527050 | Chr.7:100458543 - 100528352 on Build GRCh38 | Loss | C7orf61 TSC22D4 NYAP1 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | MIR6840 PPP1R35 MEPCE PMS2P1 STAG3L5P NYAP1 PILRB SPDYE3 ZCWPW1 C7orf61 TSC22D4 STAG3L5P-PVRIG2P-PILRB PVRIG2P PILRA |
nsv951385 | Chr.7:100467278 - 100496477 on Build GRCh38 | Deletion | TSC22D4 NYAP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |