Assay Details
Target Gene Details
Entrez Gene ID: | 84820 |
Gene Name: | RNA polymerase II subunit J4, pseudogene |
Gene Aliases: |
RPB11-phi |
Location: |
Chr.7:43940895-44019194 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR2J4 | NR_003655.3 | |||
BX537878.1 | ||||
BX538054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759527 | Chr.7:43912940 - 44017192 on Build GRCh38 | Gain | SPDYE1 POLR2J4 LOC107986791 URGCP UBE2D4 |
nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain | SPDYE1 LINC00957 MYL7 POLM RASA4CP MIR6837 PGAM2 MIR4649 POLR2J4 DBNL YKT6 POLD2 MIR6838 AEBP1 GCK |
nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain | SPDYE1 LINC00957 MYL7 POLM RASA4CP URGCP MIR6837 PGAM2 URGCP-MRPS24 MIR4649 MRPS24 POLR2J4 DBNL POLD2 LOC107986791 MIR6838 AEBP1 UBE2D4 GCK |
nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss | SPDYE1 LINC00957 POLM RASA4CP URGCP MIR6837 PGAM2 URGCP-MRPS24 MRPS24 POLR2J4 DBNL LOC107986791 MIR6838 UBE2D4 |