Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL26 | NM_006072.4 | NP_006063.1 | ||
XM_017011671.1 | XP_016867160.1 | |||
XM_017011672.1 | XP_016867161.1 | |||
AA926779.1 | ||||
AB010447.1 | BAA84579.1 | |||
AB016542.1 | BAA36704.1 | |||
AF096296.1 | AAF09265.1 | |||
AF142434.1 | AAD46495.1 | |||
AY358893.1 | AAQ89252.1 | |||
BG211121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613737 | Chr.7:75769270 - 75786583 on Build GRCh38 | Gain | CCL26 |
nsv607516 | Chr.7:75731521 - 75771091 on Build GRCh38 | Gain | HIP1 CCL26 |
dgv6468n100 | Chr.7:75701432 - 75777782 on Build GRCh38 | Gain | HIP1 CCL26 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | NSUN5P1 GTF2IP1 LOC100133091 SNORA14A LOC441259 POR LOC105375355 PMS2P3 YWHAG HSPB1 STAG3L1 SSC4D CCL26 LOC107986711 PMS2P10 GTF2IP7 STYXL1 SPDYE5 GTF2IRD2B POMZP3 MDH2 SPDYE16 TRIM73 ZP3 DTX2 PMS2P2 RHBDD2 LOC541473 CCL24 HIP1 MIR4651 POM121C FDPSP2 SRRM3 NCF1C TMEM120A |
nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain | CCL24 HIP1 CCL26 |
esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain | HIP1 CCL26 |
esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss | CCL24 HIP1 CCL26 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | LOC100133091 SNORA14A POR LOC105375355 PMS2P3 YWHAG HSPB1 SSC4D CCL26 LOC107986711 GTF2IP7 STYXL1 SPDYE5 POMZP3 MDH2 SPDYE16 ZP3 DTX2 RHBDD2 CCL24 HIP1 MIR4651 POM121C FDPSP2 SRRM3 TMEM120A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |