Assay Details
Target Gene Details
Entrez Gene ID: | 10095 |
Gene Name: | actin related protein 2/3 complex subunit 1B |
Gene Aliases: |
ARC41, p40-ARC, p41-ARC |
Location: |
Chr.7:99374675-99394801 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARPC1B | NM_005720.3 | NP_005711.1 | ||
XM_006715825.1 | XP_006715888.1 | |||
XM_006715826.1 | XP_006715889.1 | |||
AF006084.1 | AAB64189.1 | |||
AK314818.1 | ||||
AU143525.1 | ||||
BC002562.2 | AAH02562.1 | |||
BC002988.1 | AAH02988.2 | |||
BC007555.1 | AAH07555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv831068 | Chr.7:99221949 - 99408858 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv951379 | Chr.7:99375678 - 99405377 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)