Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL26 | NM_006072.4 | NP_006063.1 | ||
XM_017011672.1 | XP_016867161.1 | |||
AY358893.1 | AAQ89252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613737 | Chr.7:75769270 - 75786583 on Build GRCh38 | Gain | CCL26 |
dgv6468n100 | Chr.7:75701432 - 75777782 on Build GRCh38 | Gain | HIP1 CCL26 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | TMEM120A LOC107986711 NCF1C STAG3L1 MDH2 RHBDD2 PMS2P3 HSPB1 ZP3 LOC100133091 SNORA14A LOC541473 CCL26 LOC105375355 YWHAG STYXL1 GTF2IP1 POR CCL24 HIP1 SPDYE16 POMZP3 GTF2IP7 SPDYE5 SSC4D POM121C TRIM73 LOC441259 PMS2P2 GTF2IRD2B PMS2P10 FDPSP2 NSUN5P1 DTX2 MIR4651 SRRM3 |
esv3613738 | Chr.7:75775011 - 75778869 on Build GRCh38 | Loss | CCL26 |
nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain | CCL24 HIP1 CCL26 |
esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain | HIP1 CCL26 |
esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss | CCL24 HIP1 CCL26 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | TMEM120A LOC107986711 MDH2 RHBDD2 PMS2P3 HSPB1 ZP3 LOC100133091 SNORA14A CCL26 LOC105375355 YWHAG STYXL1 POR CCL24 HIP1 SPDYE16 POMZP3 GTF2IP7 SPDYE5 SSC4D POM121C FDPSP2 DTX2 MIR4651 SRRM3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |