Assay Details
Target Gene Details
Entrez Gene ID: | 101929580 |
Gene Name: | general transcription factor IIi pseudogene 23 |
Gene Aliases: |
- |
Location: |
Chr.7:66847450-66884273 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2IP23 | NR_135738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613520 | Chr.7:66748771 - 66896312 on Build GRCh38 | Gain | GTF2IP23 RABGEF1 GTF2IRD1P1 |
nsv981514 | Chr.7:66845152 - 66901253 on Build GRCh38 | Duplication | GTF2IP23 |
esv3891136 | Chr.7:66809968 - 67401188 on Build GRCh38 | Gain | GTF2IP23 MIR4650-1 SBDS RABGEF1 TYW1 TMEM248 LINC01372 STAG3L4 PMS2P4 GTF2IRD1P1 LOC644794 |