Assay Details
Target Gene Details
Entrez Gene ID: | 155185 |
Gene Name: | archaelysin family metallopeptidase 1 |
Gene Aliases: |
- |
Location: |
Chr.7:2679522-2767486 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMZ1 | NM_001284355.2 | NP_001271284.1 | ||
NM_001321766.1 | NP_001308695.1 | |||
NM_133463.3 | NP_597720.1 | |||
XM_011515150.2 | XP_011513452.1 | |||
XM_011515151.2 | XP_011513453.1 | |||
XM_017011774.1 | XP_016867263.1 | |||
XM_017011775.1 | XP_016867264.1 | |||
XM_017011776.1 | XP_016867265.1 | |||
AB075830.1 | ||||
AK092107.1 | ||||
AK289723.1 | ||||
BC146753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv605871 | Chr.7:2665632 - 2705490 on Build GRCh38 | Loss | AMZ1 |
nsv464257 | Chr.7:2629953 - 2705490 on Build GRCh38 | Loss | AMZ1 TTYH3 |
nsv950793 | Chr.7:2592167 - 2703366 on Build GRCh38 | Deletion | AMZ1 IQCE TTYH3 |
nsv471227 | Chr.7:2589386 - 2729230 on Build GRCh38 | Loss | AMZ1 IQCE TTYH3 GNA12 |
esv3611848 | Chr.7:2688293 - 2694228 on Build GRCh38 | Gain | AMZ1 |
dgv11144n54 | Chr.7:2620317 - 2712853 on Build GRCh38 | Loss | AMZ1 TTYH3 |
dgv401n21 | Chr.7:2600507 - 2714033 on Build GRCh38 | Loss | AMZ1 IQCE TTYH3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |