Assay Details
Target Gene Details
Entrez Gene ID: | 7589 |
Gene Name: | zinc finger and SCAN domain containing 21 |
Gene Aliases: |
NY-REN-21, ZNF38, Zipro1 |
Location: |
Chr.7:100049770-100065041 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZSCAN21 | NM_145914.2 | NP_666019.1 | ||
XM_005250568.4 | XP_005250625.1 | |||
XM_006716113.3 | XP_006716176.1 | |||
XM_017012584.1 | XP_016868073.1 | |||
XM_017012585.1 | XP_016868074.1 | |||
AF155100.1 | AAD42866.1 | |||
AK075324.1 | ||||
AK292267.1 | ||||
AK301871.1 | ||||
AK313884.1 | ||||
AL136865.1 | CAB66799.1 | |||
BC047309.1 | AAH47309.1 | |||
BI764964.1 | ||||
CR749301.1 | CAH18156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)