Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv821252 | Chr.7:76033649 - 76039546 on Build GRCh38 | Deletion |
![]() |
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3613750 | Chr.7:76031380 - 76045732 on Build GRCh38 | Loss |
![]() |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss |
![]() |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs77381667] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)