Assay Details
Target Gene Details
Entrez Gene ID: | 57224 |
Gene Name: | NHS like 1 |
Gene Aliases: |
C6orf63 |
Location: |
Chr.6:138422043-138692616 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NHSL1 | NM_001144060.1 | 8 | 4443 | NP_001137532.1 |
NM_020464.1 | 7 | 4923 | NP_065197.1 | |
XM_005267062.4 | 8 | 6707 | XP_005267119.1 | |
XM_011535965.2 | 9 | 6761 | XP_011534267.1 | |
XM_011535966.2 | 10 | 6437 | XP_011534268.1 | |
XM_011535967.2 | 10 | 6381 | XP_011534269.1 | |
XM_011535968.2 | 8 | 6629 | XP_011534270.1 | |
XM_011535969.2 | 9 | 6087 | XP_011534271.1 | |
XM_011535970.2 | 9 | 6951 | XP_011534272.1 | |
XM_011535971.2 | 8 | 5218 | XP_011534273.1 | |
XM_011535972.2 | 9 | 5032 | XP_011534274.1 | |
XM_011535973.2 | 8 | 5384 | XP_011534275.1 | |
XM_011535974.2 | 9 | 6176 | XP_011534276.1 | |
XM_011535976.1 | 9 | 4428 | XP_011534278.1 | |
XM_017011086.1 | 10 | 6347 | XP_016866575.1 | |
XM_017011087.1 | 9 | 5381 | XP_016866576.1 | |
XM_017011088.1 | 7 | 5975 | XP_016866577.1 | |
XM_017011089.1 | 8 | 4976 | XP_016866578.1 | |
AB037778.1 | 3 | 1983 | BAA92595.1 | |
AK000699.1 | 2 | 1314 | ||
AK025199.1 | 5 | 1533 | ||
AK025347.1 | 3 | 1947 | ||
AK299585.1 | 3 | 838 | ||
BX647246.1 | 4 | 3816 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752090 | Chr.6:138395100 - 138705640 on Build GRCh38 | Gain | MIR3145 HEBP2 NHSL1 FLJ46906 |