Assay Details
Target Gene Details
Entrez Gene ID: | 57224 |
Gene Name: | NHS like 1 |
Gene Aliases: |
C6orf63 |
Location: |
Chr.6:138422043-138692616 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752090 | Chr.6:138395100 - 138705640 on Build GRCh38 | Gain | FLJ46906 MIR3145 HEBP2 NHSL1 |