Assay Details
Target Gene Details
Entrez Gene ID: | 2099 |
Gene Name: | estrogen receptor 1 |
Gene Aliases: |
ER, ESR, ESRA, ESTRR, Era, NR3A1 |
Location: |
Chr.6:151654148-152103274 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ESR1 | XM_017010376.1 | XP_016865865.1 | ||
XM_017010377.1 | XP_016865866.1 | |||
XM_017010380.1 | XP_016865869.1 | |||
AB685814.1 | ||||
AB685815.1 | ||||
AB685816.1 | ||||
AB685817.1 | ||||
AB685818.1 | ||||
AB685819.1 | ||||
AB685820.1 | ||||
X86816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830844 | Chr.6:151638649 - 151860891 on Build GRCh38 | Gain |
![]() |
esv2761004 | Chr.6:151528892 - 152050641 on Build GRCh38 | Gain+Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)