Assay Details
Target Gene Details
Entrez Gene ID: | 57380 |
Gene Name: | MRS2, magnesium transporter |
Gene Aliases: |
HPT, MRS2L |
Location: |
Chr.6:24402908-24426194 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRS2 | NM_001286264.1 | NP_001273193.1 | ||
NM_001286265.1 | NP_001273194.1 | |||
NM_001286266.1 | NP_001273195.1 | |||
NM_020662.3 | NP_065713.1 | |||
NR_104423.1 | ||||
XM_005249242.1 | XP_005249299.1 | |||
AF288288.1 | AAG01170.1 | |||
AF293076.1 | AAK38615.1 | |||
AK054587.1 | ||||
AK291058.1 | ||||
AK297418.1 | ||||
AK298849.1 | ||||
AK299825.1 | ||||
BC001028.2 | AAH01028.2 | |||
BC069009.1 | AAH69009.1 | |||
BP338269.1 | ||||
DC305123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026638 | Chr.6:24347744 - 24475072 on Build GRCh38 | Gain | KAAG1 DCDC2 MRS2 GPLD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |