Assay Details
Target Gene Details
Entrez Gene ID: | 6648 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD |
Location: |
Chr.6:159679064-159762529 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain | SNORA29 WTAP LOC401281 LOC100129518 SNORA20 MRPL18 ACAT2 SOD2 TCP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |