Assay Details
Target Gene Details
Entrez Gene ID: | 83463 |
Gene Name: | MAX dimerization protein 3 |
Gene Aliases: |
BHLHC13, MAD3, MYX |
Location: |
Chr.5:177305500-177312291 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MXD3 | NM_001142935.1 | NP_001136407.1 | ||
NM_031300.3 | NP_112590.1 | |||
AF114834.1 | AAP97233.1 | |||
AK057034.1 | BAB71352.1 | |||
AK222579.1 | BAD96299.1 | |||
AK303397.1 | ||||
AK316468.1 | ||||
AL522759.3 | ||||
AL833959.1 | CAD38809.2 | |||
BC000745.2 | AAH00745.1 | |||
BC032586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509103 | Chr.5:177308751 - 177414575 on Build GRCh38 | Insertion | RGS14 SLC34A1 MXD3 PFN3 F12 LMAN2 |
nsv600364 | Chr.5:177299634 - 177432847 on Build GRCh38 | Loss | RGS14 SLC34A1 NSD1 MXD3 PFN3 PRELID1 F12 RAB24 GRK6 LMAN2 |
nsv428132 | Chr.5:177191316 - 177375443 on Build GRCh38 | Gain | RGS14 NSD1 MXD3 PRELID1 RAB24 LMAN2 |
nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication | SLC34A1 DOK3 PRELID1 F12 GRK6 LMAN2 FAM193B DBN1 RGS14 PRR7 DDX41 PDLIM7 MXD3 PFN3 PRR7-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |