Assay Details
Target Gene Details
Entrez Gene ID: | 10923 |
Gene Name: | SUB1 homolog, transcriptional regulator |
Gene Aliases: |
P15, PC4, p14 |
Location: |
Chr.5:32531778-32604079 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUB1 | XM_017008986.1 | XP_016864475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759338 | Chr.5:32245255 - 32738538 on Build GRCh38 | Loss | MTMR12 MIR579 SUB1 ZFR NPR3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |