Assay Details
Target Gene Details
Entrez Gene ID: | 10699 |
Gene Name: | corin, serine peptidase |
Gene Aliases: |
ATC2, CRN, Lrp4, PEE5, TMPRSS10 |
Location: |
Chr.4:47593998-47838106 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORIN | NM_001278585.1 | NP_001265514.1 | ||
NM_006587.3 | NP_006578.2 | |||
AF113248.1 | AAF21966.1 | |||
AF133845.1 | AAD31850.1 | |||
AK302816.1 | ||||
AK304038.1 | ||||
BC110451.2 | AAI10452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829923 | Chr.4:47434122 - 47676426 on Build GRCh38 | Gain | CORIN GABRB1 COMMD8 ATP10D MIR8053 |
dgv5202n100 | Chr.4:47486414 - 47904818 on Build GRCh38 | Gain | NFXL1 CORIN LOC101927179 ATP10D MIR8053 |
nsv1003341 | Chr.4:47605279 - 47649975 on Build GRCh38 | Gain | CORIN |
nsv594118 | Chr.4:47558369 - 47915713 on Build GRCh38 | Loss | NFXL1 CORIN LOC101927179 LOC101927157 ATP10D MIR8053 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |