Assay Details
Target Gene Details
Entrez Gene ID: | 4636 |
Gene Name: | myosin light chain 5 |
Gene Aliases: |
- |
Location: |
Chr.4:674199-682033 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYL5 | XM_006713886.2 | XP_006713949.1 | ||
XM_017008245.1 | 1 | 917 | XP_016863734.1 | |
XM_017008246.1 | 1 | 935 | XP_016863735.1 | |
XM_017008247.1 | 1 | 484 | XP_016863736.1 | |
XM_017008248.1 | XP_016863737.1 | |||
AK123974.1 | 1 | 1869 | ||
AK125529.1 | ||||
BC040050.1 | AAH40050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593206 | Chr.4:632173 - 741362 on Build GRCh38 | Loss | ATP5I PDE6B PCGF3 LOC101928521 MFSD7 MYL5 |
esv2759217 | Chr.4:609419 - 937566 on Build GRCh38 | Loss | CPLX1 ATP5I LOC107986211 PDE6B PCGF3 LOC101928521 MFSD7 MYL5 TMEM175 LOC100129917 GAK |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | SLC26A1 NKX1-1 PDE6B CRIPAK TMED11P CTBP1 CPLX1 LOC100130872 TMEM175 LOC100129917 GAK CTBP1-AS FGFRL1 LOC107986211 LOC101928521 CTBP1-AS2 UVSSA DGKQ MAEA RNF212 ATP5I PCGF3 SPON2 MFSD7 MYL5 LOC105374344 IDUA LOC101928548 |
nsv829834 | Chr.4:671688 - 781736 on Build GRCh38 | Loss | ATP5I LOC107986211 PCGF3 MFSD7 MYL5 LOC100129917 |
nsv950325 | Chr.4:620112 - 796912 on Build GRCh38 | Deletion | CPLX1 ATP5I LOC107986211 PDE6B PCGF3 LOC101928521 MFSD7 MYL5 LOC100129917 |
nsv593208 | Chr.4:674211 - 688444 on Build GRCh38 | Loss | ATP5I MFSD7 MYL5 |
nsv508986 | Chr.4:527899 - 756994 on Build GRCh38 | Insertion | ATP5I LOC107986211 PDE6B PCGF3 PIGG LOC101928521 MFSD7 MYL5 |
nsv10413 | Chr.4:673415 - 690512 on Build GRCh38 | Gain | ATP5I MFSD7 MYL5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |