Assay Details
Target Gene Details
Entrez Gene ID: | 105377590 |
Gene Name: | uncharacterized LOC105377590 |
Gene Aliases: |
- |
Location: |
Chr.4:185471431-185472263 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105377590 | NR_133944.1 | |||
AA844691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv596373 | Chr.4:185451672 - 185605031 on Build GRCh38 | Gain | SORBS2 LOC105377590 PDLIM3 CCDC110 |
dgv5493n100 | Chr.4:185446588 - 185605031 on Build GRCh38 | Gain | SORBS2 C4orf47 LOC105377590 PDLIM3 CCDC110 |
nsv1033523 | Chr.4:185394656 - 185473803 on Build GRCh38 | Gain | LRP2BP UFSP2 ANKRD37 C4orf47 LOC105377590 CCDC110 |
esv3603499 | Chr.4:185472129 - 185501859 on Build GRCh38 | Loss | LOC105377590 PDLIM3 |
esv3603496 | Chr.4:185447616 - 185603060 on Build GRCh38 | Gain | SORBS2 C4orf47 LOC105377590 PDLIM3 CCDC110 |
nsv517035 | Chr.4:185445161 - 185600889 on Build GRCh38 | Gain+Loss | SORBS2 C4orf47 LOC105377590 PDLIM3 CCDC110 |
nsv527026 | Chr.4:185404275 - 185632326 on Build GRCh38 | Gain | SORBS2 UFSP2 C4orf47 LOC105377590 PDLIM3 CCDC110 |
esv3565185 | Chr.4:185471847 - 185500852 on Build GRCh38 | Deletion | LOC105377590 PDLIM3 |
esv2759309 | Chr.4:185391971 - 185553542 on Build GRCh38 | Loss | LRP2BP UFSP2 ANKRD37 C4orf47 LOC105377590 PDLIM3 CCDC110 |
nsv428456 | Chr.4:185328666 - 185492417 on Build GRCh38 | Gain+Loss | SNX25 LRP2BP UFSP2 ANKRD37 C4orf47 LOC105377590 CCDC110 |
nsv1029738 | Chr.4:185450051 - 185515559 on Build GRCh38 | Gain | LOC105377590 PDLIM3 CCDC110 |