Assay Details
Target Gene Details
Entrez Gene ID: | 323 |
Gene Name: | amyloid beta precursor protein binding family B member 2 |
Gene Aliases: |
FE65L, FE65L1 |
Location: |
Chr.4:40810027-41216714 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10486 | Chr.4:40747595 - 40935088 on Build GRCh38 | Gain |
![]() ![]() |
nsv461346 | Chr.4:40829635 - 40855177 on Build GRCh38 | Loss |
![]() |
nsv829912 | Chr.4:40720092 - 40840600 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)