Assay Details
Target Gene Details
Entrez Gene ID: | 79730 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 7 |
Gene Aliases: |
- |
Location: |
Chr.4:40749874-40810601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN7 | NM_024677.4 | NP_078953.3 | ||
XM_017008611.1 | XP_016864100.1 | |||
XM_017008612.1 | XP_016864101.1 | |||
XM_017008613.1 | XP_016864102.1 | |||
XM_017008614.1 | XP_016864103.1 | |||
XM_017008615.1 | XP_016864104.1 | |||
AK024063.1 | BAB14808.1 | |||
AK304048.1 | ||||
BC036568.2 | AAH36568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10486 | Chr.4:40747595 - 40935088 on Build GRCh38 | Gain |
![]() ![]() |
esv3600317 | Chr.4:40677719 - 40779019 on Build GRCh38 | Gain |
![]() |
nsv525845 | Chr.4:40681959 - 40776145 on Build GRCh38 | Gain |
![]() |
nsv829912 | Chr.4:40720092 - 40840600 on Build GRCh38 | Loss |
![]() ![]() |
dgv5195n100 | Chr.4:40676645 - 40785736 on Build GRCh38 | Gain |
![]() |
nsv829911 | Chr.4:40651428 - 40806750 on Build GRCh38 | Gain |
![]() |
dgv1476e212 | Chr.4:40663647 - 40768602 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)