Assay Details
Target Gene Details
Entrez Gene ID: | 80854 |
Gene Name: | SET domain containing lysine methyltransferase 7 |
Gene Aliases: |
KMT7, SET7, SET7/9, SET9 |
Location: |
Chr.4:139495934-139556769 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SETD7 | NM_001306199.1 | NP_001293128.1 | ||
AK310856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894040 | Chr.4:139499096 - 139522362 on Build GRCh38 | Loss | SETD7 |
nsv830085 | Chr.4:139393507 - 139586168 on Build GRCh38 | Loss | LOC101927451 RAB33B LOC105377450 SETD7 |
nsv830086 | Chr.4:139494751 - 139671217 on Build GRCh38 | Gain | LOC105377622 MGST2 LOC101927490 SETD7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |