Assay Details
Target Gene Details
Entrez Gene ID: | 2549 |
Gene Name: | GRB2 associated binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.4:143336830-143474565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAB1 | NM_002039.3 | NP_002030.2 | ||
NM_207123.2 | NP_997006.1 | |||
XM_017007969.1 | XP_016863458.1 | |||
XM_017007970.1 | XP_016863459.1 | |||
XM_017007971.1 | XP_016863460.1 | |||
AK289767.1 | ||||
AK314635.1 | ||||
BC064848.1 | AAH64848.1 | |||
DA109618.1 | ||||
U43885.1 | AAC50380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026110 | Chr.4:143318672 - 143370995 on Build GRCh38 | Gain |
![]() ![]() |
nsv437941 | Chr.4:143364530 - 143374799 on Build GRCh38 | Loss |
![]() |
dgv5411n100 | Chr.4:143367285 - 143378410 on Build GRCh38 | Gain |
![]() |
nsv1018566 | Chr.4:143363774 - 143377777 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)