Assay Details
Target Gene Details
Entrez Gene ID: | 57406 |
Gene Name: | abhydrolase domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.3:58237502-58294736 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD6 | NM_001320126.1 | NP_001307055.1 | ||
NM_020676.6 | NP_065727.4 | |||
XM_005265334.3 | XP_005265391.1 | |||
XM_005265335.3 | XP_005265392.1 | |||
AF225418.1 | AAG09720.1 | |||
AK122983.1 | ||||
AK172797.1 | BAD18771.1 | |||
AK225942.1 | ||||
AK313168.1 | ||||
BC001698.2 | AAH01698.1 | |||
DA266414.1 | ||||
DB171182.1 | ||||
HY038261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834703 | Chr.3:58072791 - 58298770 on Build GRCh38 | Gain | DNASE1L3 LOC105377105 FLNB LOC105377650 ABHD6 |
nsv528147 | Chr.3:57929747 - 58384450 on Build GRCh38 | Gain | PXK DNASE1L3 LOC105377105 RPP14 FLNB LOC105377650 SLMAP ABHD6 |
nsv1001814 | Chr.3:58158881 - 58354197 on Build GRCh38 | Gain | PXK DNASE1L3 LOC105377105 RPP14 FLNB LOC105377650 ABHD6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |