Assay Details
Target Gene Details
Entrez Gene ID: | 114112 |
Gene Name: | thioredoxin reductase 3 |
Gene Aliases: |
TGR, TR2, TRXR3 |
Location: |
Chr.3:126607052-126655124 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNRD3 | NM_001173513.1 | NP_001166984.1 | ||
NM_052883.1 | NP_443115.1 | |||
AF133519.1 | AAD39929.1 | |||
AF171055.1 | AAD51325.1 | |||
AK299502.1 | ||||
BC030028.2 | AAH30028.1 | |||
BC050032.1 | AAH50032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv591524 | Chr.3:126614957 - 126669032 on Build GRCh38 | Gain | TXNRD3 NUP210P1 |
dgv4852n100 | Chr.3:126607261 - 126669480 on Build GRCh38 | Gain | TXNRD3 NUP210P1 TXNRD3NB |
nsv829718 | Chr.3:126629911 - 126809810 on Build GRCh38 | Gain | CHCHD6 TXNRD3 NUP210P1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |