Assay Details
Target Gene Details
Entrez Gene ID: | 100287362 |
Gene Name: | protease, serine 46 |
Gene Aliases: |
TESSP6 |
Location: |
Chr.3:46719583-46736431 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS46 | NM_001205271.1 | NP_001192200.1 | ||
AW183721.1 | ||||
DB058077.1 | ||||
DB099248.1 | ||||
DY654973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516623 | Chr.3:46638573 - 46811189 on Build GRCh38 | Loss | TMIE PRSS46 PRSS50 PRSS43 PRSS45 ALS2CL |
nsv590179 | Chr.3:46687169 - 46748907 on Build GRCh38 | Loss | TMIE PRSS46 PRSS50 PRSS45 ALS2CL |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss | SCAP ELP6 CSPG5 SETD2 NBEAL2 MYL3 SNORD13P3 PRSS43 CCDC12 PRSS45 NRADDP KLHL18 SMARCC1 TMIE PRSS46 PTH1R KIF9 PRSS50 PTPN23 PRSS42 KIF9-AS1 ALS2CL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |