Assay Details
Target Gene Details
Entrez Gene ID: | 100288960 |
Gene Name: | protease, serine 43 |
Gene Aliases: |
TESSP3 |
Location: |
Chr.3:46753043-46812690 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS43 | XM_017007617.1 | XP_016863106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv102e203 | Chr.3:46747685 - 46838512 on Build GRCh38 | Loss |
![]() ![]() |
dgv4734n100 | Chr.3:46751875 - 46818330 on Build GRCh38 | Gain+Loss |
![]() |
nsv1160936 | Chr.3:46807689 - 46827338 on Build GRCh38 | Deletion |
![]() |
nsv1012097 | Chr.3:46790548 - 46831502 on Build GRCh38 | Gain |
![]() ![]() |
dgv4738n100 | Chr.3:46790459 - 46827326 on Build GRCh38 | Loss |
![]() |
dgv4731n100 | Chr.3:46733893 - 46827326 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1372e212 | Chr.3:46751582 - 46818593 on Build GRCh38 | Loss |
![]() |
dgv4732n100 | Chr.3:46750043 - 46838650 on Build GRCh38 | Loss |
![]() ![]() |
dgv8308n54 | Chr.3:46811189 - 46894037 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv3798 | Chr.3:46737140 - 46824116 on Build GRCh38 | Deletion |
![]() ![]() |
dgv4737n100 | Chr.3:46770667 - 46818330 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)