Assay Details
Target Gene Details
Entrez Gene ID: | 729085 |
Gene Name: | family with sequence similarity 198 member A |
Gene Aliases: |
C3orf41 |
Location: |
Chr.3:42979267-43057715 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM198A | NM_001129908.2 | NP_001123380.2 | ||
AK091001.1 | ||||
AK311059.1 | ||||
AK311069.1 | ||||
DA262563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv590149 | Chr.3:42926105 - 43133715 on Build GRCh38 | Gain | FAM198A KRBOX1-AS1 POMGNT2 KRBOX1 |