Assay Details
Target Gene Details
Entrez Gene ID: | 100873975 |
Gene Name: | CNTN4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:3039661-3061145 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNTN4-AS1 | NR_046554.1 | |||
BM920658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1013843 | Chr.3:3058965 - 3088961 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)