Assay Details
Target Gene Details
Entrez Gene ID: | 2829 |
Gene Name: | X-C motif chemokine receptor 1 |
Gene Aliases: |
CCXCR1, GPR5 |
Location: |
Chr.3:46016990-46086803 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XCR1 | XM_017006196.1 | XP_016861685.1 | ||
XM_017006197.1 | XP_016861686.1 | |||
XM_017006198.1 | XP_016861687.1 | |||
XM_017006199.1 | XP_016861688.1 | |||
XM_017006200.1 | XP_016861689.1 | |||
XM_017006201.1 | XP_016861690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759145 | Chr.3:45974972 - 46078908 on Build GRCh38 | Loss | FYCO1 XCR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |