Assay Details
Target Gene Details
Entrez Gene ID: | 100873945 |
Gene Name: | HLTF antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:149086332-149102823 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HLTF-AS1 | NR_046648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829753 | Chr.3:148954528 - 149120758 on Build GRCh38 | Gain | GYG1 HLTF HLTF-AS1 |
nsv528849 | Chr.3:149054135 - 149117654 on Build GRCh38 | Gain | HLTF HLTF-AS1 |