Assay Details
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED12L | NM_053002.5 | NP_443728.3 | ||
XM_006713487.3 | XP_006713550.1 | |||
XM_011512390.2 | XP_011510692.1 | |||
XM_011512394.2 | XP_011510696.1 | |||
XM_011512399.2 | XP_011510701.1 | |||
XM_017005676.1 | XP_016861165.1 | |||
XM_017005677.1 | XP_016861166.1 | |||
XM_017005678.1 | XP_016861167.1 | |||
XM_017005679.1 | XP_016861168.1 | |||
XM_017005680.1 | XP_016861169.1 | |||
AF388364.1 | AAL09579.1 | |||
AF388365.1 | AAL09580.1 | |||
AF399708.1 | AAO38813.1 | |||
AK128165.1 | ||||
BC144559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759188 | Chr.3:151073838 - 151239697 on Build GRCh38 | Gain | CLRN1-AS1 GPR171 MED12L P2RY14 |
nsv829756 | Chr.3:150974849 - 151171951 on Build GRCh38 | Gain+Loss | CLRN1-AS1 MED12L |
nsv592045 | Chr.3:151052827 - 151377305 on Build GRCh38 | Gain | P2RY12 CLRN1-AS1 GPR171 GPR87 P2RY13 MED12L P2RY14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |