Assay Details
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RANBP2 | XM_017004623.1 | XP_016860112.1 |
Target Gene Details
Entrez Gene ID: | 344558 |
Gene Name: | SH3 domain containing ring finger 3 |
Gene Aliases: |
POSH2, SH3MD4 |
Location: |
Chr.2:109129439-109504636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3RF3 | NM_001099289.2 | NP_001092759.1 | ||
XM_011511109.2 | XP_011509411.1 | |||
DQ307286.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |