Assay Details
Target Gene Details
Entrez Gene ID: | 5443 |
Gene Name: | proopiomelanocortin |
Gene Aliases: |
ACTH, CLIP, LPH, MSH, NPP, POC |
Location: |
Chr.2:25160853-25168851 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POMC | NM_000939.3 | NP_000930.1 | ||
NM_001035256.2 | NP_001030333.1 | |||
NM_001319204.1 | NP_001306133.1 | |||
NM_001319205.1 | NP_001306134.1 | |||
AI341269.1 | ||||
BC065832.1 | AAH65832.1 | |||
BT019918.1 | AAV38721.1 | |||
CB988233.1 | ||||
CD106051.1 | ||||
CD108235.1 | ||||
CR541826.1 | CAG46625.1 | |||
DQ884404.1 | ||||
M38297.1 | AAA60140.1 | |||
S76492.1 | AAC60644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv57n68 | Chr.2:25041473 - 25257024 on Build GRCh38 | Loss | POMC DNMT3A LINC01381 EFR3B |
esv33372 | Chr.2:25064335 - 25385947 on Build GRCh38 | Loss | MIR1301 DTNB POMC DNMT3A LINC01381 EFR3B |
nsv1008732 | Chr.2:24907673 - 25349661 on Build GRCh38 | Loss | MIR1301 DNAJC27 POMC DNAJC27-AS1 DNMT3A ADCY3 LINC01381 EFR3B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |