Assay Details
Target Gene Details
Entrez Gene ID: | 151258 |
Gene Name: | solute carrier family 38 member 11 |
Gene Aliases: |
AVT2 |
Location: |
Chr.2:164897745-164955721 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC38A11 | NM_001199148.1 | NP_001186077.1 | ||
NM_173512.2 | NP_775783.1 | |||
XM_005246350.4 | XP_005246407.1 | |||
XM_006712337.3 | XP_006712400.1 | |||
XM_011510737.2 | XP_011509039.1 | |||
XM_011510742.2 | XP_011509044.1 | |||
XM_017003455.1 | XP_016858944.1 | |||
XM_017003456.1 | XP_016858945.1 | |||
XM_017003457.1 | XP_016858946.1 | |||
XM_017003458.1 | XP_016858947.1 | |||
XM_017003459.1 | XP_016858948.1 | |||
XM_017003460.1 | XP_016858949.1 | |||
XM_017003461.1 | XP_016858950.1 | |||
XM_017003463.1 | XP_016858952.1 | |||
XM_017003464.1 | XP_016858953.1 | |||
XM_017003465.1 | XP_016858954.1 | |||
AK097141.1 | BAC04962.1 | |||
AK293998.1 | ||||
BC125157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834444 | Chr.2:164865781 - 165038205 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)