Assay Details
Target Gene Details
Entrez Gene ID: | 100271715 |
Gene Name: | Rho guanine nucleotide exchange factor 33 |
Gene Aliases: |
- |
Location: |
Chr.2:38919363-38975449 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF33 | NM_001145451.2 | NP_001138923.2 | ||
DV080278.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |