Assay Details
Target Gene Details
Entrez Gene ID: | 5538 |
Gene Name: | palmitoyl-protein thioesterase 1 |
Gene Aliases: |
CLN1, INCL, PPT |
Location: |
Chr.1:40072710-40097470 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPT1 | NM_000310.3 | NP_000301.1 | ||
NM_001142604.1 | NP_001136076.1 | |||
XM_005271008.1 | XP_005271065.1 | |||
AK075496.1 | ||||
AK301682.1 | ||||
AK302232.1 | ||||
AK310799.1 | ||||
AK312287.1 | ||||
BC008426.1 | AAH08426.1 | |||
CR542053.1 | CAG46850.1 | |||
U44772.1 | AAB06236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829660 | Chr.1:40073032 - 40266408 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
esv2750836 | Chr.1:40038107 - 40093576 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)