Assay Details
Target Gene Details
Entrez Gene ID: | 148979 |
Gene Name: | GLIS family zinc finger 1 |
Gene Aliases: |
- |
Location: |
Chr.1:53506233-53739171 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLIS1 | NM_147193.2 | NP_671726.2 | ||
XM_017000408.1 | XP_016855897.1 | |||
XM_017000409.1 | XP_016855898.1 | |||
XM_017000410.1 | XP_016855899.1 | |||
XM_017000411.1 | XP_016855900.1 | |||
AK090634.1 | BAC03494.1 | |||
AK093474.1 | BAC04178.1 | |||
BC101799.1 | ||||
BC104911.1 | AAI04912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522165 | Chr.1:53506376 - 53539387 on Build GRCh38 | Loss | GLIS1 |
nsv428443 | Chr.1:53452879 - 53563563 on Build GRCh38 | Gain | GLIS1 DMRTB1 |
nsv509269 | Chr.1:53296636 - 53583096 on Build GRCh38 | Insertion | GLIS1 DMRTB1 LRP8 SLC25A3P1 LOC105378732 |
nsv829882 | Chr.1:53449992 - 53629940 on Build GRCh38 | Loss | GLIS1 DMRTB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |