Assay Details
Target Gene Details
Entrez Gene ID: | 391013 |
Gene Name: | phospholipase A2 group IIC |
Gene Aliases: |
- |
Location: |
Chr.1:20163085-20177364 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G2C | NM_001316722.1 | NP_001303651.1 | ||
XM_017001270.1 | XP_016856759.1 | |||
AW135717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain | MUL1 MIR6084 VWA5B1 UBXN10-AS1 PLA2G5 CAMK2N1 PLA2G2C PINK1-AS HP1BP3 SH2D5 PLA2G2D CDA PINK1 PLA2G2F LINC01141 FAM43B UBXN10 KIF17 DDOST |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |