Assay Details
Target Gene Details
Entrez Gene ID: | 5226 |
Gene Name: | phosphogluconate dehydrogenase |
Gene Aliases: |
6PGD |
Location: |
Chr.1:10398992-10420511 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PGD | NM_001304451.1 | NP_001291380.1 | ||
NM_001304452.1 | NP_001291381.1 | |||
NM_002631.3 | NP_002622.2 | |||
AK290404.1 | ||||
AK296889.1 | ||||
AK298830.1 | ||||
AK300952.1 | ||||
AK303898.1 | ||||
AK304423.1 | ||||
BC000368.2 | AAH00368.1 | |||
U30255.1 | AAA75302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain | APITD1 APITD1-CORT KIF1B PGD PEX14 CORT DFFA |
esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss | APITD1 APITD1-CORT KIF1B PGD PEX14 CORT MIR1273D DFFA UBE4B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |