Assay Details
Target Gene Details
Entrez Gene ID: | 84838 |
Gene Name: | zinc finger protein 496 |
Gene Aliases: |
NIZP1, ZFP496, ZKSCAN17, ZSCAN49 |
Location: |
Chr.1:247297413-247333376 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833137 | Chr.1:247274015 - 247444126 on Build GRCh38 | Gain | ZNF496 LOC107985115 NLRP3 |
esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss | OR2L8 OR2W3 OR2B11 OR2T8 MIR3916 ZNF670-ZNF695 ZNF669 OR9H1P ZNF670 OR2L13 OR2W5 OR6F1 OR2M5 C1orf229 OR13G1 OR2AK2 ZNF124 OR2M1P GCSAML VN1R5 OR2L2 OR14A16 OR2C3 OR2L1P AHCTF1 ZNF695 LOC107985115 OR2L3 OR2G3 ZNF496 OR1C1 NLRP3 TRIM58 OR2G2 OR2L5 GCSAML-AS1 OR11L1 |
esv3589514 | Chr.1:247297392 - 247354590 on Build GRCh38 | Gain | ZNF496 LOC107985115 |
nsv509790 | Chr.1:247311000 - 247445876 on Build GRCh38 | Insertion | ZNF496 LOC107985115 NLRP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |