Assay Details
Target Gene Details
Entrez Gene ID: | 65220 |
Gene Name: | NAD kinase |
Gene Aliases: |
dJ283E3.1 |
Location: |
Chr.1:1751232-1780514 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NADK | NM_001198993.1 | NP_001185922.1 | ||
NM_001198994.1 | NP_001185923.1 | |||
NM_023018.4 | NP_075394.3 | |||
XM_005244778.2 | XP_005244835.1 | |||
XM_006710837.2 | XP_006710900.1 | |||
XM_006710838.2 | XP_006710901.1 | |||
XM_006710839.1 | XP_006710902.1 | |||
XM_011542006.1 | XP_011540308.1 | |||
XM_011542007.1 | XP_011540309.1 | |||
XM_011542008.1 | XP_011540310.1 | |||
XM_011542009.2 | XP_011540311.1 | |||
XM_017002143.1 | XP_016857632.1 | |||
XM_017002145.1 | XP_016857634.1 | |||
XM_017002146.1 | XP_016857635.1 | |||
AF250320.1 | AAG44568.1 | |||
AK023114.1 | BAB14412.1 | |||
AK290161.1 | ||||
AK291599.1 | ||||
AK291728.1 | ||||
BC001709.1 | AAH01709.1 | |||
CN349189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5n111 | Chr.1:1575421 - 1823403 on Build GRCh38 | Deletion | GNB1 MMP23A SLC35E2B SLC35E2 CDK11A CDK11B FNDC10 MIB2 NADK MMP23B |
nsv834091 | Chr.1:1701359 - 1880638 on Build GRCh38 | Loss | GNB1 MMP23A SLC35E2 CDK11A NADK |
nsv545041 | Chr.1:1753033 - 1780259 on Build GRCh38 | Loss | NADK |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss | LOC105378948 MIR200A ANKRD65 CDK11B TTLL10-AS1 FNDC10 SAMD11 ATAD3A TNFRSF18 C1orf159 TNFRSF4 MIR6726 CPSF3L MXRA8 LOC100287934 SSU72 RNF223 LOC107985728 FAM132A MIR200B FAM87B UBE2J2 HES4 MMP23A CPTP LOC284600 LINC00115 SCNN1D LOC107985729 MIB2 ACAP3 LINC01342 ISG15 LOC100288069 ATAD3C SDF4 CDK11A DVL1 PUSL1 LOC100130417 AGRN LOC148413 PLEKHN1 ATAD3B SLC35E2B FAM41C LOC102724312 TMEM240 SLC35E2 TAS1R3 LOC100288175 KLHL17 TTLL10 LINC01128 TMEM88B MMP23B MIR429 MRPL20 PERM1 NOC2L CCNL2 MIR6727 MIR6808 NADK B3GALT6 AURKAIP1 VWA1 |
nsv1160797 | Chr.1:1566216 - 1805918 on Build GRCh38 | Duplication | GNB1 MMP23A SLC35E2B SLC35E2 CDK11A CDK11B FNDC10 MIB2 NADK MMP23B SSU72 |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss | LOC105378948 DDX11L1 MIR200A ANKRD65 TTLL10-AS1 FNDC10 SAMD11 ATAD3A GABRD CPSF3L LOC100129534 MXRA8 RNF223 LOC107985728 LOC100134822 FAAP20 MIR6859-1 LOC100133331 UBE2J2 MMP23A CPTP LOC101928626 LOC107985729 ACAP3 TMEM52 OR4F5 LOC105378589 WASH7P ISG15 LOC100288069 OR4F29 MIR6723 LOC100132287 CDK11A DVL1 AGRN SLC35E2B FAM41C FAM138A TTLL10 LOC105378949 LINC01128 PERM1 NOC2L MORN1 CCNL2 LOC107984841 MIR6727 MIR6808 NADK B3GALT6 OR4F16 CDK11B TNFRSF18 C1orf159 TNFRSF4 MIR6726 SKI MTND1P23 LOC100287934 SSU72 LOC729737 CFAP74 FAM132A LOC105378947 LOC105378591 MIR200B FAM87B MIR6859-2 PRKCZ HES4 LOC284600 LINC00115 SCNN1D MIB2 LOC102725121 GNB1 LOC105378592 LINC01342 LOC100506504 ATAD3C SDF4 MIR1302-2 PUSL1 LOC100130417 CALML6 LOC148413 PLEKHN1 ATAD3B LOC102724312 TMEM240 SLC35E2 TAS1R3 LOC100288175 KLHL17 TMEM88B MMP23B MTND2P28 MIR429 MRPL20 AURKAIP1 VWA1 |
nsv1160926 | Chr.1:1772699 - 1905488 on Build GRCh38 | Duplication | GNB1 LOC105378949 NADK |
esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss | LOC105378589 GNB1 ANKRD65 CDK11B FNDC10 ATAD3A GABRD ATAD3C CDK11A SSU72 CALML6 LOC148413 ATAD3B CFAP74 SLC35E2B LOC102724312 TMEM240 SLC35E2 LOC105378591 LOC105378949 TMEM88B MMP23B PRKCZ MRPL20 MMP23A CCNL2 LOC107985729 MIB2 NADK TMEM52 VWA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |